Orthophosphoric acid 99% ≥99%, EMSURE® analytical reagent, Supelco®

Supplier: Merck
Danger

Synonyms: Phosphoric acid

M-Clarity™ quality level = MQ300

1.00565.0500
1.00565.0500EA 84.5 GBP
1.00565.0500
Orthophosphoric acid 99% ≥99%, EMSURE® analytical reagent, Supelco®
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 3453
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Solubility 750 - 850 g/l (20 °C)
Melting Point 42 °C
Molar Mass 97.99 g/mol
Boiling Point 213 °C (1013 hPa)
Vapor Pressure 2 hPa (20 °C)
Density 1.87 g/cm3 (25 °C)
pH  - 0.5 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR