Dodecamolybdophosphoric acid hydrate, EMSURE® ACS, Reag. Ph. Eur. analysis_for, Supelco®

Supplier: Merck
Danger

Synonyms: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate

MQ300

1.00532.0025 1.00532.0100
1.00532.0025EA 138 GBP
1.00532.0025 1.00532.0100
Dodecamolybdophosphoric acid hydrate, EMSURE® ACS, Reag. Ph. Eur. analysis_for, Supelco®
Dodecamolybdophosphoric acid hydrate
Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
MW: 1843.27 g/mol
Melting Pt: 78…90 °C
Density: 1.62 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
EINECS: 234-713-5
UN: 3260
ADR: 8 II
Merck Index: 12,07498

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 78 - 90 °C (anhydrous substance)
Molar Mass 1825.28 g/mol (anhydrous substance)
Bulk Density 980 kg/m3
pH (H2O) strongly acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR