Hydrochloric acid 32%, Supelco®

Supplier: Merck
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

MQ200

1.00313.2500 1.00313.9025 1.00313.9180
1.00313.2500EA 49.7 GBP
1.00313.2500 1.00313.9025 1.00313.9180
Hydrochloric acid 32%, Supelco®
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -40 °C
Boiling Point 83 - 90 °C
Vapor Pressure 21.3 hPa (20 °C)
Density 1.16 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR