Orthoboric acid, EMSURE® Reag. Ph. Eur., ISO analysis_for, Supelco®

Supplier: Merck
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

MQ300

1.00165.0100 1.00165.0500 1.00165.1000 1.00165.5000 1.00165.9025
1.00165.0100EA 37.2 GBP
1.00165.0100 1.00165.0500 1.00165.1000 1.00165.5000 1.00165.9025
Orthoboric acid, EMSURE® Reag. Ph. Eur., ISO analysis_for, Supelco®
ortho-Boric acid
Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185
Melting Pt: 160
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 46.5 g/l (20 °C)
Melting Point 171 °C (decomposition)
Molar Mass 61.83 g/mol
Bulk Density 400 - 600 kg/m3
Vapor Pressure 2.7 hPa (20 °C)
Density 1.44 g/cm3 (20 °C)
pH 3.8 - 4.8 (33 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR