Acetic acid glacial_AcOH, anhydrous 100% Alkalimetric, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®

Supplier: Merck
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

MQ300

1.00063.1000 1.00063.1011 1.00063.2500 1.00063.2511 1.00063.9026 1.00063.9200 1.00063.2510 1.00063.0510 1.00063.1010
1.00063.1000EA 28.04 GBP
1.00063.1000 1.00063.1011 1.00063.2500 1.00063.2511 1.00063.9026 1.00063.9200 1.00063.2510 1.00063.0510 1.00063.1010
Acetic acid glacial_AcOH, anhydrous 100% Alkalimetric, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3) II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Alkalimetric) ≥99.8%
Identity Passes Test
Color ≤10 Hazen
Acetaldehyde ≤2 ppm
Acetic Anhydride ≤100 ppm
Titratable Base ≤0.0004 meq/g
Solidification Temperature ≥16.3 °C
Chloride (Cl) ≤0.4 ppm
Phosphate (PO₄) ≤0.4 ppm
Heavy metals (as Pb) ≤0.5 ppm
Sulfate (SO₄) ≤0.4 ppm
Ag (Silver) ≤0.005 ppm
Al (Aluminium) ≤0.020 ppm
As (Arsenic) ≤0.010 ppm
Au (Gold) ≤0.010 ppm
B (Boron) ≤0.100 ppm
Ba (Barium) ≤0.010 ppm
Be (Beryllium) ≤0.005 ppm
Bi (Bismuth) ≤0.050 ppm
Ca (Calcium) ≤0.100 ppm
Cd (Cadmium) ≤0.020 ppm
Co (Cobalt) ≤0.010 ppm
Cr (Chromium) ≤0.020 ppm
Cu (Copper) ≤0.010 ppm
Fe (Iron) ≤0.050 ppm
Ga (Gallium) ≤0.050 ppm
Ge (Germanium) ≤0.020 ppm
Hg (Mercury) ≤0.005 ppm
In (Indium) ≤0.050 ppm
K (Potassium) ≤0.100 ppm
Li (Lithium) ≤0.010 ppm
Mg (Magnesium) ≤0.050 ppm
Mn (Manganese) ≤0.010 ppm
Mo (Molybdenum) ≤0.010 ppm
Na (Sodium) ≤0.200 ppm
Ni (Nickel) ≤0.020 ppm
Pb (Lead) ≤0.010 ppm
Pt (Platinum) ≤0.100 ppm
Sn (Tin) ≤0.050 ppm
Sr (Strontium) ≤0.010 ppm
Ti (Titanium) ≤0.050 ppm
Tl (Thallium) ≤0.020 ppm
V (Vanadium) ≤0.010 ppm
Zn (Zinc) ≤0.030 ppm
Zr (Zirconium) ≤0.050 ppm
Substances Reducing Potassium Dichromate Passes Test
Substances Reducing Potassium Permanganate ≤20 ppm
Evaporation Residue ≤5 ppm
Water ≤0.2
Dilution test Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR