2-Heptanone, Supelco®

Supplier: Merck
Danger


MQ100

02476-1ML
02476-1MLEA 75 GBP
02476-1ML
2-Heptanone, Supelco®
2-Heptanone
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 150
Melting Pt: –35 °C
Density: 0.8111 g/cm³ (20 °C)
Flash Pt: 43
Storage Temperature: Ambient
MDL Number: MFCD00009513
CAS Number: 110-43-0
EINECS: 203-767-1
UN: 1110
ADR: 3 III
Merck Index: 13,04683
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d2c208d2bc918d894235d8ea8280c79cf6822c5&vid=4cc42ebe1e55429eb4a2a612c8ba49f0757fed82&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR