tri-Sodium citrate dihydrate 99, reagent

Supplier: VWR Chemicals

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

0101-1KGEA 141 GBP
0101-1KG 0101-2.5KG 0101-500G 0101-12KG 0101-50KG
tri-Sodium citrate dihydrate 99, reagent
tri-Sodium citrate dihydrate

A biological buffer used for cell culture, in vitro, enzyme assays and some electrophoretic applications at physiological pH.

Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium ≤0.005%
Chloride ≤0.005%
DNase none detected
Heavy Metals (as Pb) ≤0.001%
Insolubles ≤0.005%
Iron ≤0.0005%
Loss on Drying (%) 10.0 – 13.0
pH (5%, Water) @25 °C 7.0 – 9.0
Protease none detected
Purity (Dry Basis) (%) ≥99%
RNase none detected
Sulphate ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR