Copper ≥99.5% (metals basis), powder -40/+100 mesh

Supplier: Thermo Fisher Scientific
Warning

00908.36 00908.A3
00908.36EA 61.3 GBP
00908.36 00908.A3
Copper ≥99.5% (metals basis), powder -40/+100 mesh
Copper

Purity: 99.5% CAS No.: 7440-50-8 Odour: Odourless UN No.: 3077 MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3077
ADR: 9 III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR