cAMP and cGMP conjugates
Supplier: Biotium
00020-1 00020 00021-1 00021 00022-1 00022
00020-1EA 596 GBP
00020-1 00020. 00021-1 00021. 00022-1 00022.
cAMP and cGMP conjugates
Assays Immunological Assays
Conjugates for probing cAMP receptors or measuring cGMP in immunoassays.
  • Biotin-cAMP can be used to probe cAMP receptors
  • Biotin-cGMP has been used as a tracer in immunoassays for cGMP measurement in plasma or urine samples
  • Cyanine 644-cAMP can be used to probe cAMP receptors
Biotin-cAMP, diisopropylethylammonium salt is a biotin-conjugated cAMP analogue that can be used to probe cAMP receptors.
Biotin-cGMP, diisopropylethylammonium salt has been used as a tracer in immunoassays for the cGMP measurement in plasma or urine samples. Cyanine 644-cAMP is a fluorescently labeled cAMP analogue and can be used to probe cAMP receptors.
Cyanine 644-cAMP λExEm (MeOH) = 644/670 nm. Cyanine 644-cAMP is a fluorescently labeled cAMP analog and can be used to probe cAMP receptors. Single molecules of fluorescently labeled cAMP have been imaged as the probe binds to the receptor on the surface of Dictyostelium cells.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c71b89f5bb9fc271307593b4b8ef5e991a2f04b3&vid=d7793f85ad046d34aaa668c3d44b1f4f310e1852&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82c2abf8119929b0d22bc20795e78d2ec8e3bbf8&vid=6c7db50eb305c92289a7e89511ddffe90c6fd077&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ea0d4df8385fb9502b1d494a8939666151f73dc&vid=7de9ea46793766c29757b0730c51db4f55b1f190&prid=web&clid=SAPDAM
Order Now
SPECIFICATIONS
Environmentally Preferable