Analytical Grade Reagents
A range of high purity reagents suitable for a variety of analytical techniques. These bulk packs offer extensive guaranteed specifications, with potential contaminents meaured on every batch of material to ensure a consistent performance time after time. The quoted limits represent 'maximum permitted levels of impurities' not 'typical analyses'.