Acridine orange (hemizinc salt) for electrophoresis
Supplier: VWR Chemicals
|
Danger
|
Synonyms:
Acridine orange hemi(Zinc chloride) salt
, Acridine orange
, Basic orange 14
, 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt
, Euchrysine 3RX
, Euchrysin GNY
, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride
, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt)
A fluorescent stain for nucleic acids in agarose and polyacrylamide gels. Concentrations of 120 ?M will detect DNA at levels of 25-50 ng per band. Also used for to localize nucleic acids in a variety of cell types
Formula:
C₁₇H₂₀ClN₃·HCl·½ZnCl₂ MW: 369,96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00081043 CAS Number: 10127-02-3 EINECS: 233-353-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...