Auramine O, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloridePyoctanunum aureum4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloridePyoktanin YellowBasic Yellow 2Canary Yellow

1.01301.0050EA 0 GBP
1.01301.0000 1.01301.0050
Auramine O, Sigma-Aldrich®
Auramine O
Formula: C₁₇H₂₂ClN₃
MW: 303,84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012484
CAS Number: 2465-27-2
EINECS: 219-567-2
UN: 3077
ADR: 9,III
IVDLogo

Order Now

Specification Test Results

Solubility 10 g/l (20 °C)
Melting Point 265 °C (decomposition)
Molar Mass 303.84 g/mol
Bulk Density 430 kg/m3
pH 6 - 7 (10 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR