Methyl green zinc chloride salt

Supplier: Molekula
Warning

Synonyms: CI 42590Methyl green zinc chloride double saltMethyl GreenEthyl Green

MOLEM31140436EA 0 GBP
MOLEM31140436
Methyl green zinc chloride salt
Methyl Green zinc chloride salt
Formula: C₂₇H₃₅BrClN₃·xZnCl₂
MW: 653,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151094
CAS Number: 7114-03-6
EINECS: 230-415-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR