Ampicillin sodium salt
Supplier: LGC Standards PROMOCHEM
|
Danger
|
Synonyms:
D(-)-α-Aminobenzylpenicillin Sodium Salt
PROOMM0253.00EA
775
GBP
PROOMM0253.00
Ampicillin sodium salt
Ampicillin sodium salt
Formula:
C₁₆H₁₈N₃NaO₄S MW: 371,39 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00064313 CAS Number: 69-52-3 EINECS: 200-708-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...