Ampicillin sodium salt

Supplier: LGC Standards PROMOCHEM
Danger

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

PROOICRS0212EA 283 GBP
PROOICRS0212
Ampicillin sodium salt
Ampicillin sodium salt
Formula: C₁₆H₁₈N₃NaO₄S
MW: 371,39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
EINECS: 200-708-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR