Pentachloropyridine for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
8.41070.0025EA
75
GBP
8.41070.0025
Pentachloropyridine for synthesis, Sigma-Aldrich®
Pentachloropyridine
Formula:
C₅Cl₅N MW: 251,33 g/mol Boiling Pt: 279…280 °C (1013 hPa) Melting Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006230 CAS Number: 2176-62-7 EINECS: 218-535-5 UN: 3077 ADR: 9,III |
Specification Test Results
Solubility | 0.009 g/l (25 °C) |
Melting Point | 124 - 126 °C |
Molar Mass | 251.32 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...