Pentachloropyridine ≥98%

Supplier: Thermo Scientific
Danger

A13842.22EA 56.4 GBP
A13842.22 A13842.36 A13842.0E
Pentachloropyridine ≥98%
Pentachloropyridine
Formula: C₅Cl₅N
MW: 251,33 g/mol
Boiling Pt: 279…280 °C (1013 hPa)
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00006230
CAS Number: 2176-62-7
EINECS: 218-535-5
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR