Erythrosin B disodium salt, Sigma-Aldrich®
Supplier: SIGMA-ALDRICH MICROSCOPY
|
Warning
|
Synonyms:
FD&C Red #3
, Erythrosin 239
, Sodium 2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)
, Erythrosine 3B
, Erythrosine sodium
, C.I. 45430
, Erythrosin B
, Acid Red 51
M-Clarity™ quality level = MQ200
198269-25GEA
52.5
GBP
198269-25G
Erythrosin B disodium salt, Sigma-Aldrich®
Erythrosin B disodium salt
Formula:
C₂₀H₆I₄Na₂O₅ MW: 879,86 g/mol |
MDL Number:
MFCD00144257 CAS Number: 16423-68-0 EINECS: 240-474-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...