Carbazole
Supplier: LGC Standards PROMOCHEM
|
Warning
|
PROOERM-FC021EA
350
GBP
PROOERM-FC021
Carbazole
Carbazole
Formula:
C₁₂H₉N MW: 167,21 g/mol Boiling Pt: 355 °C (1013 hPa) Melting Pt: 236 °C Density: 1,26 g/cm³ (20 °C) |
MDL Number:
MFCD00004960 CAS Number: 86-74-8 EINECS: 201-696-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...