Nuclear Fast Red

Supplier: RAL (REACTIFS RAL SA)

Synonyms: Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonateCI 60760Helio Fast Rubin BBL

720-1193EA 186 GBP
720-1193
Nuclear Fast Red
Nuclear Fast Red
Formula: C₁₄H₈NNaO₇S
MW: 357,28 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00078493
CAS Number: 6409-77-4
EINECS: 229-088-0
Merck Index: 13,06512
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR