Ammonium iron (III) citrate (14,5 - 16,0% Fe), green, powder FCC, J.T.Baker®
Supplier: Avantor
Synonyms:
Ammonium iron (III) citrate
, Ferric Ammonium Citrate
, Ammonium ferric citrate
1977-01EA
128
GBP
1977-01
Ammonium iron (III) citrate (14,5 - 16,0% Fe), green, powder FCC, J.T.Baker®
Ammonium iron (III) citrate
Formula:
C₆H₅+4y)Fe(x)N(y)O₇ |
MDL Number:
MFCD00013099 CAS Number: 1185-57-5 EINECS: 214-686-6 Merck Index: 13,00518 |
Specification Test Results
Meets F.C.C. Requirements | |
Food GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
FCC - Assay (as Fe) | 14.5 - 16.0 % |
FCC - Identification | Passes Test |
FCC - Ferric Citrate | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Mercury (Hg) | ≤1 mg/kg |
FCC - Oxalate (C₂O₄) | Passes Test |
FCC - Sulfate (SO₄) | ≤0.3 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...