Gemfibrozil

Supplier: LGC Standards PROMOCHEM

Synonyms: 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeric acidCI-7195-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid

PROOMM0641.00EA 1270 GBP
PROOMM0641.00
Gemfibrozil
Gemfibrozil
Formula: C₁₅H₂₂O₃
MW: 250,34 g/mol
Boiling Pt: 395 °C (1013 hPa)
Melting Pt: 59…61 °C
MDL Number: MFCD00079335
CAS Number: 25812-30-0
EINECS: 247-280-2
Merck Index: 12,04394

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR