Rigid polyethylene tags (B-145) are printable, non-adhesive, tear-resistant, flame retardent tags used for a wide array of identification applications.
- Constructed from a 9,5 mil, cross laminated polyethylene film and a thermal transfer printable topcoat
- Suitable for thermal transfer printing, then folded over and held together in a two-ply format with a pressure sensitive adhesive with tear-resistant qualities
- Operating temperature of −40 to +100 °C (−40 to +212 °F)
- Matte finish
This multipurpose material can be used for a variety of tag applications including asset tracking, component and equipment labelling, data and telecommunications labelling, facility identification, general identification, inventory and inspection labelling, safety identification, and wire and cable labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6210.
Certifications: RoHS compliant.
Packaging: 100 tags per box.