3,4-Dicyanoaniline 97%
Supplier: Apollo Scientific
Synonyms:
4-Aminophthalonitrile
, 3,4-Dicyanoaniline
APOSOR953694-25GEA
197
GBP
APOSOR953694-25G
APOSOR953694-5G
APOSOR953694-100G
3,4-Dicyanoaniline 97%
3,4-Dicyanoaniline
Formula:
C₈H₅N₃ MW: 143,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041741 CAS Number: 56765-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...