4-Aminophenol

Supplier: Apollo Scientific
Warning

Synonyms: 4-Hydroxyaniline

APOSOR8828-100GEA 72.2 GBP
APOSOR8828-100G
4-Aminophenol
4-Aminophenol
Formula: H₂NC₆H₄OH
MW: 109,13 g/mol
Boiling Pt: 284 °C
Melting Pt: 185…190 °C
Density: 1,29 g/cm³ (25 °C)
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00007869
CAS Number: 123-30-8
EINECS: 204-616-2
UN: 2512
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR