Ochratoxin A (from Aspergillus ochraceus)
Supplier: Apollo Scientific
|
Danger
|
APOSBIO1001-5MGEA
479
GBP
APOSBIO1001-5MG
APOSBIO1001-1MG
Ochratoxin A (from Aspergillus ochraceus)
Ochratoxin A
Mycotoxin.
Formula:
C₂₀H₁₈ClNO₆ MW: 403,8 g/mol Melting Pt: 105…110 °C Storage Temperature: Freezer |
MDL Number:
MFCD00078079 CAS Number: 303-47-9 EINECS: 206-143-7 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...