Dextran, M.W. 40, BioChemica

Supplier: Apollo Scientific
Warning

APOSBID1013-500GEA 302 GBP
APOSBID1013-500G APOSBID1013-100G
Dextran, M.W. 40, BioChemica
Dextran
Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR