RALA361510
Supplier: RAL (REACTIFS RAL SA)
720-1165EA 41.7 GBP
720-1165
RALA361510
Used in Gram-Hücker R Kit To differentiate Gram-positive from Gram-negative Bacteria.
Used in Gram-Hücker R Kit Ref 361520
Gram-Hücker staining is based on the same principle than the Gram-Nicolle staining. The structure of the bacterial walls and their permeability to alcohol help differentiate the Gram-negative bacteria from the Gram-positive ones. In this technique, Carbolic Gentian Violet is replaced by Crystal Violet Oxalate (that does not require filtration) and Ziehl-Carbolic Fuchsin by Safranin.