Dansyl chloride
Supplier: Molekula
|
Danger
|
Synonyms:
5-(Dimethylamino)naphthalene-1-sulphonyl chloride
MOLE22561258-5GEA
263
GBP
MOLE22561258-5G
Dansyl chloride
Dansyl chloride
Formula:
C₁₂H₁₂ClNO₂S MW: 269,75 g/mol Melting Pt: 69…73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003985 CAS Number: 605-65-2 EINECS: 210-092-6 UN: 3261 ADR: 8,III Merck Index: 13,02840 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...