(R)-(+)-1-Phenylethyl isocyanate 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
(R)-(+)-α-Phenethyl isocyanate, (R)-(+)-1-Phenethyl isocyanate, (R)-(+)-1-phenylethylisocyanate, (R)-(+)-α-Methylbenzyl isocyanate
205320050
ACRO205320050EA
270
GBP
ACRO205320050
(R)-(+)-1-Phenylethyl isocyanate 98%
(R)-(+)-1-Phenylethyl isocyanate
Formula:
C₉H₉NO Boiling Pt: 55…56 °C (3,3hPa) Density: 1,05 g/cm³ Flash Pt: 65 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06739387 CAS Number: 33375-06-3 EINECS: 251-485-2 UN: 2206 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5110 to 1.5150 (20°C, 589 nm) |
GC | ≥97.5 % |
Specific optical rotation | +9° to +11° (20°C, 589 nm) (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...