Lanthanum(III) chloride hydrate ACS

Supplier: Thermo Scientific
Danger

Synonyms: Lanthanum trichloride hydrate

ACRO413010500EA 108 GBP
ACRO413010500
Lanthanum(III) chloride hydrate ACS
Lanthanum (III) chloride hydrate
Formula: Cl₃La·H₂O
MW: 263,28 g/mol
Boiling Pt: ∼860 °C (1013 hPa)
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00149753
CAS Number: 20211-76-1
EINECS: 233-237-5
UN: 1759

Order Now

Specification Test Results

Appearance White Adhering crystalline powder or crystals
Assay 64.5 to 70.0 % (LaCl3)
Calcium (Ca) ≤10 ppm
Magnesium (Mg) ≤10 ppm
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR