Neutral red, Gurr™ for microscopical staining

Supplier: VWR Chemicals

Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochlorideToluylene redCI 500403-Amino-7-dimethylamino-2-methylphenazineN8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochlorideBasic Red 5

340564AEA 193 GBP
340564A 340565B
Neutral red, Gurr™ for microscopical staining
Neutral red
Formula: C₁₅H₁₇ClN₄
MW: 288,78 g/mol
MDL Number: MFCD00012651
CAS Number: 553-24-2
IVDLogo

Order Now

Specification Test Results

Dye content (spectrophotometrically) Min. 90 %
Identity (UV/VIS-Spectrum) Passes test
Absorption max. Lambda max (Ethanol 50%) 540 --> 543 nm
Spec.Absorptiv.(Lm;0.005g/l;ethanol 50%) 1420 --> 1610
TLC-Test Passes test
Loss on drying (110°C) Max. 12 %
Suitability for microscopy Passes test
Basophilic granules Brick red
Eosinophilic granules Yellow or light orange
Mitochondria Green
Neutrophilic granules Pale pink
Vacuoles of monocytes Salmon

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR