Nicotinic acid (Niacin)
Supplier: LGC Standards PROMOCHEM
|
Warning
|
Synonyms:
Pyridine-3-carboxylic acid
, 3-Pyridinecarboxylic acid
, Vitamin B3
, 3-Picolinic acid
, Nicotinic acid
, Niacin
PROOICRS0358EA
283
GBP
PROOICRS0358
Nicotinic acid (Niacin)
Nicotinic acid (Niacin)
Formula:
C₆H₅NO₂ MW: 123,11 g/mol Boiling Pt: 258 °C (1013 hPa) Melting Pt: 142 °C Density: 1,473 g/cm³ (20 °C) Flash Pt: 193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006391 CAS Number: 59-67-6 EINECS: 200-441-0 Merck Index: 12,06612 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...