Cyanogen bromide

Supplier: Molekula
Danger

MOLE16432166-100GEA 142 GBP
MOLE16432166-100G
Cyanogen bromide
Cyanogen bromide
Formula: CBrN
MW: 105,92 g/mol
Boiling Pt: 61,4 °C (1013 hPa)
Melting Pt: 52 °C
Density: 2,06 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011597
CAS Number: 506-68-3
EINECS: 208-051-2
UN: 1889
ADR: 6.1,I
Merck Index: 12,02763

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR