4-Aminophenol ≥99%, purified
Supplier: VWR Chemicals
|
Warning
|
Synonyms:
4-Hydroxyaniline
21168.262EA
70.6
GBP
21168.262
4-Aminophenol ≥99%, purified
4-Aminophenol
Formula:
H₂NC₆H₄OH MW: 109,13 g/mol Boiling Pt: 284 °C (1013 hPa) Melting Pt: ∼186 °C Density: 1,3 g/cm³ (20 °C) Flash Pt: 195 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00007869 CAS Number: 123-30-8 EINECS: 204-616-2 UN: 2512 ADR: 6.1,III Merck Index: 13,00461 |
Specification Test Results
Assay | Min. 99 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...