Charcoal activated, powder USP, J.T.Baker®

Supplier: Avantor

Synonyms: Activated carbonCarbon activatedActivated charcoal

1560-01EA 0 GBP
1560-01
Charcoal activated, powder USP, J.T.Baker®
Charcoal activated
Formula: C
MW: 12,01 g/mol
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Microbial Limits - Escherichia coli Passes Test
Microbial Limits - Salmonella Passes Test
Reaction Passes Test
USP - Loss on Drying ≤15.0 %
Residue on Ignition ≤4.0 %
Acid-Soluble Substances ≤3.5 %
Chloride (Cl) ≤0.2 %
USP - Sulfate (SO₄) ≤0.2 %
USP - Sulfide (S) Passes Test
USP - Cyanogen Compounds Passes Test
USP - Uncarbonized Constituents Passes Test
Adsorptive Power: Alkaloids Passes Test
Adsorptive Power: Dyes Passes Test
Typical Particle Size, % thru 150 micron ≥97 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR