Anti-Ubiquitin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Enzo Life Sciences
ENZOBMLPW09350100EA 380 GBP
ALEXBMLPW09350100 ENZOBMLPW09350100 ENZOBMLPW09350025 ALEXBMLPW09350025
Anti-Ubiquitin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Ubiquitin modification of cellular proteins has been shown to be involved with a wide range of biochemical processes including proteasomal degradation, signal transduction, DNA repair, endocytosis and autophagy.

  • For Western Blot

Recognizes mono- and poly-ubiquitin protein conjugates, free polyubiquitin chains and free ubiquitin.

Type: Primary
Antigen: UBIQUITIN
Clonality: Monoclonal
Clone:
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR