Anti-MDA Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ALEXALX210879R100EA 888 GBP
ALEXALX210879R100 ENZOALX210879R100
Anti-MDA Rabbit Polyclonal Antibody
Antibodies
MDA (Malondialdehyde) is a highly reactive three carbon dialdehyde produced as a byproduct of polyunsaturated fatty acid peroxidation and arachidonic acid metabolism. MDA readily combines with several functional groups on molecules including proteins, lipoproteins, and DNA. MDA modified proteins may show altered physico-chemical behavior and antigenicity. Antibodies to MDA will help to visualize the MDA-adducts.

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes MDA adducts.

Reconstitute with 100μl water.

Type: Primary
Antigen: MDA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR