Ochratoxin A 99+%
Supplier: Thermo Scientific
|
Danger
|
ACRO227400050EA
727
GBP
ACRO227400050
Ochratoxin A 99+%
Ochratoxin A
Formula:
C₂₀H₁₈ClNO₆ MW: 403,8 g/mol Melting Pt: 169 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00078079 CAS Number: 303-47-9 UN: 3462 ADR: 6.1,II |
Specification Test Results
Appearance | White to off-white Powder |
HPLC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...