Lithium bromide 4 M in THF, AcroSeal™

Supplier: Thermo Scientific
Danger

390971000.EA 497 GBP
390971000. 390978000.
Lithium bromide 4 M in THF, AcroSeal™
Lithium bromide
Formula: LiBr
MW: 86,84 g/mol
Boiling Pt: 1265 °C (1013 hPa)
Melting Pt: 550 °C
Density: 1,18 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00011077
CAS Number: 7550-35-8
UN: 2056
ADR: 3,II

Order Now

Specification Test Results

Appearance yellow to pink Solution
Titration Argentometric 29 to 31 % (LiBr via Br)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR