Rottlerin 95+%

Supplier: Thermo Scientific
Warning

ACRO328490100EA 170 GBP
ACRO328490100
Rottlerin 95+%
Rottlerin

Appearance: Orange to brown Powder

Formula: C₃₀H₂₈O₈
MW: 516,546 g/mol
Melting Pt: >200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017361
CAS Number: 82-08-6

Order Now

Specification Test Results

Appearance Orange to brown Powder
Infrared spectrum Conforms
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR