Anti-UCP3 Rabbit Polyclonal Antibody

Supplier: Oncogene
662048-100EA 877 GBP
662048-100
Anti-UCP3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UCP3 (uncoupling protein 3 (mitochondrial, proton carrier))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR