Anti-EIF2AK2 Rabbit Polyclonal Antibody

Supplier: Oncogene
527460-10EA 897 GBP
527460-10
Anti-EIF2AK2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EIF2AK2 (eukaryotic translation initiation factor 2-alpha kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR