Anti-Glial Fibrillary Acidic Protein (2.2B10) Rat Monoclonal Antibody

Supplier: Oncogene
345860-100EA 775 GBP
345860-100
Anti-Glial Fibrillary Acidic Protein (2.2B10) Rat Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Glial Fibrillary Acidic Protein (2.2B10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR