Anti-Factor H Goat Polyclonal Antibody

Supplier: Oncogene
341276-1EA 241 GBP
341276-1
Anti-Factor H Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Factor H
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR