Anti-EGFR Rabbit Polyclonal Antibody

Supplier: Oncogene
324867-10EA 861 GBP
324867-10
Anti-EGFR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR