Anti-COL7 Rabbit Polyclonal Antibody

Supplier: Oncogene
234192-500EA 694 GBP
234192-500
Anti-COL7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COL7 (collagen, type VII)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR