Anti-CD44 (A020) Rat Monoclonal Antibody [clone: A020]

Supplier: Oncogene
217594-100EA 769 GBP
217594-100
Anti-CD44 (A020) Rat Monoclonal Antibody [clone: A020]
Antibodies
Type: Primary
Antigen: CD44 (A020)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR