Anti-PFKFB4 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB137785-100EA 1100 GBP
ABCAAB137785-100
Anti-PFKFB4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PFKFB4 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR