Anti-BirA Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB106159-100EA 1090 GBP
ABCAAB106159-100
Anti-BirA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BirA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR